missing translation for 'onlineSavingsMsg'
Learn More

4-n-Octylaniline 97.0+%, TCI America™

Catalog No. O01865G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
O01865G 5 g
O018625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. O01865G Supplier TCI America Supplier No. O01865G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 16245-79-7
Molecular Formula C14H23N
Molecular Weight (g/mol) 205.35
MDL Number MFCD00007929
InChI Key ORKQJTBYQZITLA-UHFFFAOYSA-N
Synonym 4-n-octylaniline, p-octylaniline, benzenamine, 4-octyl, 4-octylphenylamine, unii-o9fn212wzz, o9fn212wzz, p-n-octylaniline, 4-octyl aniline, acmc-209dof, 4-octylaniline
PubChem CID 85346
IUPAC Name 4-octylaniline
SMILES CCCCCCCCC1=CC=C(N)C=C1

Specifications

Boiling Point 311°C
Quantity 5 g
Formula Weight 205.35
Percent Purity ≥97.0% (GC,T)
Physical Form Liquid
Chemical Name or Material 4-n-Octylaniline
EINECSNumber (3)-0206
TSCA No