missing translation for 'onlineSavingsMsg'
Learn More

4-n-Octylaniline, 99%

Catalog No. AAH2671306
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH2671306 5 g
AAH2671303 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH2671306 Supplier Thermo Scientific Chemicals Supplier No. H2671306
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 16245-79-7 | C14H23N | 205.35 g/mol

4-Octylaniline can be been used as an extractant during the extraction of microgram level concentrations of ruthenium(IV) from halide medium. It can also been used in the synthesis of selective benzimidazole based analogs of sphingosine-1-phosphate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Octylaniline can be been used as an extractant during the extraction of microgram level concentrations of ruthenium(IV) from halide medium. It can also been used in the synthesis of selective benzimidazole based analogs of sphingosine-1-phosphate.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.

Chemical Identifiers

CAS 16245-79-7
Molecular Formula C14H23N
Molecular Weight (g/mol) 205.35
MDL Number MFCD00007929
InChI Key ORKQJTBYQZITLA-UHFFFAOYSA-N
Synonym 4-n-octylaniline, p-octylaniline, benzenamine, 4-octyl, 4-octylphenylamine, unii-o9fn212wzz, o9fn212wzz, p-n-octylaniline, 4-octyl aniline, acmc-209dof, 4-octylaniline
PubChem CID 85346
IUPAC Name 4-octylaniline
SMILES CCCCCCCCC1=CC=C(N)C=C1

Specifications

Density 0.898
Boiling Point 175°C (13 mmHg)
Flash Point 110°C (230°F)
Refractive Index 1.516
Quantity 5 g
Solubility Information Insoluble in water.
Formula Weight 205.34
Percent Purity 99%
Chemical Name or Material 4-n-Octylaniline
Hazard Category H302+H312
Hazard Statement GHS H Statement
H301-H311
Toxic if swallowed.
Toxic in contact with skin.
Precautionary Statement P264b-P270-P280-P301+P312-P302+P352-P312-P330-P363-P501c
EINECSNumber 240-358-7
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only