missing translation for 'onlineSavingsMsg'
Learn More

4'-n-Octylacetophenone 97.0+%, TCI America™

Catalog No. p-7134238
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
O02075G 5 g
O020725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. O02075G Supplier TCI America Supplier No. O02075G
Only null left

Chemical Identifiers

CAS 10541-56-7
Molecular Formula C16H24O
Molecular Weight (g/mol) 232.37
MDL Number MFCD00043682
InChI Key GARQDIVXKVBJFP-UHFFFAOYSA-N
Synonym 1-4-octylphenyl ethanone, p-octylacetophenone, 4'-n-octylacetophenone, 4'-octylacetophenone, ethanone, 1-4-octylphenyl, p-n-octylacetophenone, 4-n-octylacetophenone, acetophenone, 4'-octyl, 1-acetyl-4-octylbenzene, p-octylaceto-phenone
PubChem CID 66344
IUPAC Name 1-(4-octylphenyl)ethan-1-one
SMILES CCCCCCCCC1=CC=C(C=C1)C(C)=O

Specifications

Color White-Yellow
Quantity 5 g
Formula Weight 232.37
Percent Purity ≥97.0% (GC)
Physical Form Crystalline Lumps
Chemical Name or Material 4′-n-Octylacetophenone
EINECSNumber (3)-3540
TSCA Yes