missing translation for 'onlineSavingsMsg'
Learn More

4-n-Octyl-2-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)thiophene 96.0+%, TCI America™

Catalog No. O04115G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
O04115G 5 g
O04111G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. O04115G Supplier TCI America Supplier No. O04115G

Chemical Identifiers

CAS 405165-12-0
Molecular Formula C18H31BO2S
Molecular Weight (g/mol) 322.31
MDL Number MFCD27976835
InChI Key RKYWQDDPNVTJJQ-UHFFFAOYSA-N
Synonym 4,4,5,5-Tetramethyl-2-(4-n-octyl-2-thienyl)-1,3,2-dioxaborolan, 4-n-Octyl-2-thiopheneboronic Acid Pinacol Ester
PubChem CID 25259825
IUPAC Name 4,4,5,5-tetramethyl-2-(4-octylthiophen-2-yl)-1,3,2-dioxaborolane
SMILES CCCCCCCCC1=CSC(=C1)B1OC(C)(C)C(C)(C)O1

Specifications

Color Yellow
Quantity 5 g
Formula Weight 322.31
Percent Purity ≥96.0% (GC)
Physical Form Liquid
Chemical Name or Material 4-n-Octyl-2-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)thiophene
TSCA No
Recommended Storage Refrigerator