missing translation for 'onlineSavingsMsg'
Learn More

4-n-Octanoylbiphenyl 97.0+%, TCI America™

Catalog No. O021410G
Change view
Click to view available options
Quantity:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
O021410G 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. O021410G Supplier TCI America Supplier No. O021410G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 47162-00-5
Molecular Formula C20H24O
Molecular Weight (g/mol) 280.411
MDL Number MFCD00143358
InChI Key JSNGEMRLSPQOOF-UHFFFAOYSA-N
PubChem CID 607296
IUPAC Name 1-(4-phenylphenyl)octan-1-one
SMILES CCCCCCCC(=O)C1=CC=C(C=C1)C2=CC=CC=C2

Specifications

Melting Point 100°C
Color White
Boiling Point 200°C
Quantity 10 g
Formula Weight 280.41
Percent Purity ≥97.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4-n-Octanoylbiphenyl