missing translation for 'onlineSavingsMsg'
Learn More

4-(N,N-Diphenylamino)benzaldehyde 98.0+%, TCI America™

Catalog No. D21725G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D21725G 5 g
D217225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D21725G Supplier TCI America Supplier No. D21725G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 4181-05-9
Molecular Formula C19H15NO
Molecular Weight (g/mol) 273.34
MDL Number MFCD00145131
InChI Key UESSERYYFWCTBU-UHFFFAOYSA-N
Synonym 4-diphenylamino benzaldehyde, 4-n,n-diphenylamino benzaldehyde, 4-diphenylaminobenzaldehyde, 4-diphenylamino-benzaldehyde, benzaldehyde, 4-diphenylamino, 4-formyltriphenylamine, p-formyltriphenylamine, 4-n-phenylanilino benzaldehyde, p-diphenylaminobenzaldehyde, 4-n,n-diphenylamino-benzaldehyde
PubChem CID 77846
IUPAC Name 4-(N-phenylanilino)benzaldehyde
SMILES C1=CC=C(C=C1)N(C2=CC=CC=C2)C3=CC=C(C=C3)C=O

Specifications

Melting Point 132°C
Quantity 5 g
Formula Weight 273.34
Percent Purity ≥98.0% (GC,N)
Physical Form Crystalline Powder
Chemical Name or Material 4-(N,N-Diphenylamino)benzaldehyde