missing translation for 'onlineSavingsMsg'
Learn More

4-(N,N-Dimethylaminosulfonyl)-7-amino-2,1,3-benzoxadiazole 98.0+%, TCI America™

Catalog No. D3830100MG
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D3830100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D3830100MG Supplier TCI America Supplier No. D3830100MG
Only null left

Chemical Identifiers

CAS 147611-83-4
Molecular Formula C8H10N4O3S
Molecular Weight (g/mol) 242.253
InChI Key ZRHMJRAHRITJBK-UHFFFAOYSA-N
PubChem CID 44629982
IUPAC Name 7-amino-N,N-dimethyl-2,1,3-benzoxadiazole-4-sulfonamide
SMILES CN(C)S(=O)(=O)C1=CC=C(C2=NON=C12)N

Specifications

Color Yellow
Quantity 100 mg
Formula Weight 242.25
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material 4-(N,N-Dimethylaminosulfonyl)-7-amino-2,1,3-benzoxadiazole
TSCA No
Recommended Storage Refrigerator