missing translation for 'onlineSavingsMsg'
Learn More

4-n-Heptyloxybenzaldehyde, 97%

Catalog No. AAL0896606
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0896606 5 g
AAL0896614 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0896606 Supplier Thermo Scientific Chemicals Supplier No. L0896606
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 27893-41-0 | C14H20O2 | 220.312 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 27893-41-0
Molecular Formula C14H20O2
Molecular Weight (g/mol) 220.312
MDL Number MFCD00016616
InChI Key YBCKMIZXHKVONZ-UHFFFAOYSA-N
Synonym 4-heptyloxybenzaldehyde, 4-heptyloxy benzaldehyde, p-heptyloxybenzaldehyde, 4-n-heptyloxybenzaldehyde, benzaldehyde, 4-heptyloxy, p-heptyloxy benzaldehyde, 4-n-heptoxybenzaldehyde, 4-heptyloxy-benzaldehyde, p-n-heptoxybenzaldehyde, acmc-209gzj
PubChem CID 119740
IUPAC Name 4-heptoxybenzaldehyde
SMILES CCCCCCCOC1=CC=C(C=C1)C=O

Specifications

Density 0.99
Boiling Point 152°C to 154°C (1 mmHg)
Refractive Index 1.525
Quantity 5 g
Beilstein 1912690
Sensitivity Air Sensitive
Formula Weight 220.31
Percent Purity 97%
Chemical Name or Material 4-n-Heptyloxybenzaldehyde
EINECSNumber 248-714-3
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only