missing translation for 'onlineSavingsMsg'
Learn More

4-n-Heptylaniline, 98%

Catalog No. AAH2679306
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH2679306 5 g
AAH2679314 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH2679306 Supplier Thermo Scientific Chemicals Supplier No. H2679306

CAS: 37529-27-4 | C13H21N | 191.32 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 37529-27-4
Molecular Formula C13H21N
Molecular Weight (g/mol) 191.32
MDL Number MFCD00007928
InChI Key BNEWZYZRLNNWNR-UHFFFAOYSA-N
Synonym p-heptylaniline, 4-n-heptylaniline, benzenamine, 4-heptyl, 4-heptylphenylamine, acmc-209ito, 4-heptylaniline
PubChem CID 142169
IUPAC Name 4-heptylaniline
SMILES CCCCCCCC1=CC=C(N)C=C1

Specifications

Density 0.91
Boiling Point 159°C (18 mmHg)
Flash Point >110°C (230°F)
Refractive Index 1.52
Quantity 5 g
Sensitivity Light sensitive
Formula Weight 191.31
Percent Purity 98%
Chemical Name or Material 4-n-Heptylaniline
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H301-H315-H319
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only