missing translation for 'onlineSavingsMsg'
Learn More

4-n-Dodecyloxybenzaldehyde, 98%

Catalog No. AAL0888903
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0888903 1 g
AAL0888906 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0888903 Supplier Thermo Scientific Chemicals Supplier No. L0888903
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 24083-19-0 | C19H30O2 | 290.447 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 24083-19-0
Molecular Formula C19H30O2
Molecular Weight (g/mol) 290.447
MDL Number MFCD00043523
InChI Key ZBEGLEYBWGNZJA-UHFFFAOYSA-N
Synonym p-dodecyloxybenzaldehyde, 4-dodecyloxy benzaldehyde, 4-dodecyloxybenzaldehyde, 4-n-dodecyloxybenzaldehyde, p-n-dodecoxy benzaldehyde, p-dodecyloxy benzaldehyde, 4-dodecyloxy-benzaldehyde, p-lauryloxy benzaldehyde, 1-4-formylphenoxy dodecane
PubChem CID 141069
IUPAC Name 4-dodecoxybenzaldehyde
SMILES CCCCCCCCCCCCOC1=CC=C(C=C1)C=O

Specifications

Melting Point 26°C to 29°C
Boiling Point 165°C (0.005 mmHg)
Quantity 1 g
Beilstein 2535031
Sensitivity Air Sensitive
Formula Weight 290.45
Percent Purity 98%
Chemical Name or Material 4-n-Dodecyloxybenzaldehyde
Hazard Category H315-H319-H335
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only