Learn More
4-n-Decylbenzoyl chloride, 98%
CAS: 54256-43-8 | C17H25ClO | 280.836 g/mol
Supplier: Thermo Scientific Chemicals A1723406
Description
Employed as a pharmaceutical and chemical intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsEmployed as a pharmaceutical and chemical intermediate.
Solubility
Reacts with water.
Notes
Moisture Sensitive. Store away from oxidizing agents, water/moisture. Store under inert gas.
Specifications
| 4-n-Decylbenzoyl chloride | |
| 171°C to 173°C (2 mmHg) | |
| C17H25ClO | |
| 5 g | |
| Moisture sensitive | |
| Reacts with water. | |
| CCCCCCCCCCC1=CC=C(C=C1)C(=O)Cl | |
| 280.836 | |
| 280.84 |
| 54256-43-8 | |
| >110°C (230°F) | |
| MFCD00041724 | |
| UN3265 | |
| 4-n-decylbenzoyl chloride, benzoyl chloride, 4-decyl, p-decylbenzoyl chloride, 4-decylbenzoylchloride, p-n-decylbenzoyl chloride, benzoyl chloride,4-decyl, 4-n-decyl benzoyl chloride, 4-decylbenzoic acid chloride | |
| HEWLDHSFOQXCSI-UHFFFAOYSA-N | |
| 4-decylbenzoyl chloride | |
| 104714 | |
| 98% |
Chemical Identifiers
| 54256-43-8 | |
| 280.836 | |
| HEWLDHSFOQXCSI-UHFFFAOYSA-N | |
| 4-decylbenzoyl chloride |
| C17H25ClO | |
| MFCD00041724 | |
| 104714 | |
| CCCCCCCCCCC1=CC=C(C=C1)C(=O)Cl |
Safety and Handling
GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
P210-P235-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P370+P378q-P501c
H227-H315-H319-H335
DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.
EINECSNumber : 259-046-7
TSCA : Yes
Recommended Storage : Ambient temperatures; Store under Nitrogen
RUO – Research Use Only