missing translation for 'onlineSavingsMsg'
Learn More

4-n-Butylbenzoyl chloride, 98%

CAS: 28788-62-7 | C11H13ClO | 196.67 g/mol

$346.00 - $346.00

Chemical Identifiers

CAS 28788-62-7
Molecular Formula C11H13ClO
Molecular Weight (g/mol) 196.67
MDL Number MFCD00000699
InChI Key OUOWCSJYDCPVDM-UHFFFAOYSA-N
Synonym 4-n-butylbenzoyl chloride, benzoyl chloride, 4-butyl, p-butylbenzoyl chloride, 4-n-butyl-benzoyl chloride, 4-butylbenzoylchloride, 4-n-butylbenzoylchloride, p-n-butyl benzoylchloride, p-n-butylbenzoyl chloride, 4-1-butylbenzoyl chloride, acmc-209h4o
PubChem CID 120049
IUPAC Name 4-butylbenzoyl chloride
SMILES CCCCC1=CC=C(C=C1)C(Cl)=O
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAA1508214
View Documents
Thermo Scientific Chemicals
A1508214
25 g
Each for $346.00
Only null left
 
Description

Description

4-Butylbenzoyl chloride was used in the synthesis of 2,5-bis[(4-butylbenzoyl)oxy]styrene (BBOS, monomer).

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Butylbenzoyl chloride was used in the synthesis of 2,5-bis[(4-butylbenzoyl)oxy]styrene (BBOS, monomer).

Solubility
Reacts with water.

Notes
Moisture Sensitive, store away from moisture. Store in cool, dry place in tightly closed container. Store away from oxidizing agent. Store under inert gas.
Specifications

Chemical Identifiers

28788-62-7
196.67
OUOWCSJYDCPVDM-UHFFFAOYSA-N
120049
CCCCC1=CC=C(C=C1)C(Cl)=O
C11H13ClO
MFCD00000699
4-n-butylbenzoyl chloride, benzoyl chloride, 4-butyl, p-butylbenzoyl chloride, 4-n-butyl-benzoyl chloride, 4-butylbenzoylchloride, 4-n-butylbenzoylchloride, p-n-butyl benzoylchloride, p-n-butylbenzoyl chloride, 4-1-butylbenzoyl chloride, acmc-209h4o
4-butylbenzoyl chloride

Specifications

28788-62-7
145°C to 147°C (11 mmHg)
C11H13ClO
MFCD00000699
UN3265
4-n-butylbenzoyl chloride, benzoyl chloride, 4-butyl, p-butylbenzoyl chloride, 4-n-butyl-benzoyl chloride, 4-butylbenzoylchloride, 4-n-butylbenzoylchloride, p-n-butyl benzoylchloride, p-n-butylbenzoyl chloride, 4-1-butylbenzoyl chloride, acmc-209h4o
OUOWCSJYDCPVDM-UHFFFAOYSA-N
4-butylbenzoyl chloride
120049
98%
1.051
>110°C (230°F)
1.5351
25 g
Moisture sensitive
Reacts with water.
CCCCC1=CC=C(C=C1)C(Cl)=O
196.67
196.68
4-n-Butylbenzoyl chloride
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.

P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c

H314

DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.

EINECSNumber : 249-224-2

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only