missing translation for 'onlineSavingsMsg'
Learn More

4-n-Butylbenzoyl chloride, 98%

Catalog No. AAA1508214
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1508214 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAA1508214 Supplier Thermo Scientific Chemicals Supplier No. A1508214
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 28788-62-7 | C11H13ClO | 196.67 g/mol

4-Butylbenzoyl chloride was used in the synthesis of 2,5-bis[(4-butylbenzoyl)oxy]styrene (BBOS, monomer).

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Butylbenzoyl chloride was used in the synthesis of 2,5-bis[(4-butylbenzoyl)oxy]styrene (BBOS, monomer).

Solubility
Reacts with water.

Notes
Moisture Sensitive, store away from moisture. Store in cool, dry place in tightly closed container. Store away from oxidizing agent. Store under inert gas.

Chemical Identifiers

CAS 28788-62-7
Molecular Formula C11H13ClO
Molecular Weight (g/mol) 196.67
MDL Number MFCD00000699
InChI Key OUOWCSJYDCPVDM-UHFFFAOYSA-N
Synonym 4-n-butylbenzoyl chloride, benzoyl chloride, 4-butyl, p-butylbenzoyl chloride, 4-n-butyl-benzoyl chloride, 4-butylbenzoylchloride, 4-n-butylbenzoylchloride, p-n-butyl benzoylchloride, p-n-butylbenzoyl chloride, 4-1-butylbenzoyl chloride, acmc-209h4o
PubChem CID 120049
IUPAC Name 4-butylbenzoyl chloride
SMILES CCCCC1=CC=C(C=C1)C(Cl)=O

Specifications

Density 1.051
Boiling Point 145°C to 147°C (11 mmHg)
Flash Point >110°C (230°F)
Refractive Index 1.5351
Quantity 25 g
UN Number UN3265
Sensitivity Moisture sensitive
Solubility Information Reacts with water.
Formula Weight 196.68
Percent Purity 98%
Chemical Name or Material 4-n-Butylbenzoyl chloride
Show More Show Less
Hazard Category H314
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.
EINECSNumber 249-224-2
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only