Learn More
4-n-Butylbenzenesulfonyl chloride, 97%
CAS: 54997-92-1 | C10H13ClO2S | 232.72 g/mol
$144.23 - $522.53
Chemical Identifiers
| CAS | 54997-92-1 |
|---|---|
| Molecular Formula | C10H13ClO2S |
| Molecular Weight (g/mol) | 232.72 |
| MDL Number | MFCD00173908 |
| InChI Key | OVFZELSNOHIDEF-UHFFFAOYSA-N |
| Synonym | 4-butylbenzene-1-sulfonyl chloride, 4-n-butylbenzenesulfonyl chloride, 4-n-butyl benzenesulphonyl chloride, 4-butyl-benzenesulfonyl chloride, 4-n-butylphenylsulfonyl chloride, 4-n-butylbenzenesulphonyl chloride, benzenesulfonyl chloride, 4-butyl, 4-butylphenyl chlorosulfone, pubchem5484, 4-n-butylbenzenesulfonylchloride |
| PubChem CID | 2737352 |
| IUPAC Name | 4-butylbenzenesulfonyl chloride |
| SMILES | CCCCC1=CC=C(C=C1)S(Cl)(=O)=O |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAA1657706
|
Thermo Scientific Chemicals
A1657706 |
5 g |
Each for $144.23
|
|
|||||
|
AAA1657714
|
Thermo Scientific Chemicals
A1657714 |
25 g |
Each for $522.53
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 54997-92-1 | |
| 232.72 | |
| OVFZELSNOHIDEF-UHFFFAOYSA-N | |
| 2737352 | |
| CCCCC1=CC=C(C=C1)S(Cl)(=O)=O |
| C10H13ClO2S | |
| MFCD00173908 | |
| 4-butylbenzene-1-sulfonyl chloride, 4-n-butylbenzenesulfonyl chloride, 4-n-butyl benzenesulphonyl chloride, 4-butyl-benzenesulfonyl chloride, 4-n-butylphenylsulfonyl chloride, 4-n-butylbenzenesulphonyl chloride, benzenesulfonyl chloride, 4-butyl, 4-butylphenyl chlorosulfone, pubchem5484, 4-n-butylbenzenesulfonylchloride | |
| 4-butylbenzenesulfonyl chloride |
Specifications
| 54997-92-1 | |
| 126°C to 128°C (0.6 mmHg) | |
| MFCD00173908 | |
| UN3265 | |
| 4-butylbenzene-1-sulfonyl chloride, 4-n-butylbenzenesulfonyl chloride, 4-n-butyl benzenesulphonyl chloride, 4-butyl-benzenesulfonyl chloride, 4-n-butylphenylsulfonyl chloride, 4-n-butylbenzenesulphonyl chloride, benzenesulfonyl chloride, 4-butyl, 4-butylphenyl chlorosulfone, pubchem5484, 4-n-butylbenzenesulfonylchloride | |
| CCCCC1=CC=C(C=C1)S(Cl)(=O)=O | |
| 232.72 | |
| 232.73 | |
| 4-n-Butylbenzenesulfonyl chloride |
| 13°C to 15°C | |
| C10H13ClO2S | |
| 5 g | |
| Moisture sensitive | |
| OVFZELSNOHIDEF-UHFFFAOYSA-N | |
| 4-butylbenzenesulfonyl chloride | |
| 2737352 | |
| 97% |
Safety and Handling
GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
H314-H335
DOTInformation : Hazard Class: 8; Packaging Group: III
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only