missing translation for 'onlineSavingsMsg'
Learn More

4-n-Butylbenzenesulfonyl chloride, 97%

Catalog No. AAA1657706
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1657706 5 g
AAA1657714 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1657706 Supplier Thermo Scientific Chemicals Supplier No. A1657706

CAS: 54997-92-1 | C10H13ClO2S | 232.72 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 54997-92-1
Molecular Formula C10H13ClO2S
Molecular Weight (g/mol) 232.72
MDL Number MFCD00173908
InChI Key OVFZELSNOHIDEF-UHFFFAOYSA-N
Synonym 4-butylbenzene-1-sulfonyl chloride, 4-n-butylbenzenesulfonyl chloride, 4-n-butyl benzenesulphonyl chloride, 4-butyl-benzenesulfonyl chloride, 4-n-butylphenylsulfonyl chloride, 4-n-butylbenzenesulphonyl chloride, benzenesulfonyl chloride, 4-butyl, 4-butylphenyl chlorosulfone, pubchem5484, 4-n-butylbenzenesulfonylchloride
PubChem CID 2737352
IUPAC Name 4-butylbenzenesulfonyl chloride
SMILES CCCCC1=CC=C(C=C1)S(Cl)(=O)=O

Specifications

Melting Point 13°C to 15°C
Boiling Point 126°C to 128°C (0.6 mmHg)
Quantity 5 g
UN Number UN3265
Sensitivity Moisture sensitive
Formula Weight 232.73
Percent Purity 97%
Chemical Name or Material 4-n-Butylbenzenesulfonyl chloride
Hazard Category H314-H335
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Hazard Class: 8; Packaging Group: III
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only