missing translation for 'onlineSavingsMsg'
Learn More

4-n-Butylbenzamide, 97%

Catalog No. AAA1951603
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1951603 1 g
AAA1951606 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1951603 Supplier Thermo Scientific Chemicals Supplier No. A1951603
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 107377-07-1 | C11H15NO | 177.247 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 107377-07-1
Molecular Formula C11H15NO
Molecular Weight (g/mol) 177.247
MDL Number MFCD00221466
InChI Key KFINRIKJBULSTD-UHFFFAOYSA-N
Synonym 4-n-butylbenzamide, benzamide, 4-butyl, p-butylbenzamid, acmc-20e7n2
PubChem CID 2801458
IUPAC Name 4-butylbenzamide
SMILES CCCCC1=CC=C(C=C1)C(=O)N

Specifications

Melting Point 127°C to 129°C
Quantity 1 g
Formula Weight 177.25
Percent Purity 97%
Chemical Name or Material 4-n-Butylbenzamide
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only