missing translation for 'onlineSavingsMsg'
Learn More

4'-n-Butylacetophenone, 97%

Catalog No. AAA1529606
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1529606 5 g
AAA1529614 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1529606 Supplier Thermo Scientific Chemicals Supplier No. A1529606
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 37920-25-5 | C12H16O | 176.26 g/mol

4'-n-Butylacetophenone is used as an organic chemical synthesis intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4′-n-Butylacetophenone is used as an organic chemical synthesis intermediate.

Solubility
Not miscible in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents, acids and bases.

Chemical Identifiers

CAS 37920-25-5
Molecular Formula C12H16O
Molecular Weight (g/mol) 176.26
MDL Number MFCD00017500
InChI Key MQESVSITPLILCO-UHFFFAOYSA-N
Synonym 4'-butylacetophenone, 1-4-butylphenyl ethanone, ethanone, 1-4-butylphenyl, p-butylacetophenone, 1-4-butylphenyl ethan-1-one, p-n-butylacetophenone, 4'-n-butylacetophenone, unii-6738cf9z8v, 1-acetyl-4-butylbenzene, 1-4-butyl-phenyl ethanone
PubChem CID 123470
IUPAC Name 1-(4-butylphenyl)ethanone
SMILES CCCCC1=CC=C(C=C1)C(C)=O

Specifications

Density 0.957
Boiling Point 101°C to 102°C (1.5 mmHg)
Flash Point >110°C (230°F)
Quantity 5 g
Solubility Information Not miscible in water.
Formula Weight 176.26
Percent Purity 97%
Chemical Name or Material 4'-n-Butylacetophenone
Hazard Category H302-H315-H319-H335
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
EINECSNumber 253-715-7
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only