missing translation for 'onlineSavingsMsg'
Learn More

4-n-Butoxyphenyl isocyanate, 98%

Catalog No. AAL1287706
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1287706 5 g
AAL1287703 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1287706 Supplier Thermo Scientific Chemicals Supplier No. L1287706

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 28439-86-3 | C11H13NO2 | 191.23 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 28439-86-3
Molecular Formula C11H13NO2
Molecular Weight (g/mol) 191.23
MDL Number MFCD00037042
InChI Key ASFIDVOMDQBCNP-UHFFFAOYSA-N
Synonym 4-n-butoxyphenyl isocyanate, 4-butoxyphenyl isocyanate, benzene, 1-butoxy-4-isocyanato, 1-butoxy-4-isocyanato-benzene, pubchem16232, 4-butoxybenzenisocyanate
PubChem CID 2769629
IUPAC Name 1-butoxy-4-isocyanatobenzene
SMILES CCCCOC1=CC=C(C=C1)N=C=O

Specifications

Density 1.055
Boiling Point 149°C to 150°C (3 mmHg)
Flash Point >110°C (230°F)
Refractive Index 1.522
Quantity 5 g
UN Number UN2206
Beilstein 2107851
Sensitivity Moisture sensitive
Formula Weight 191.23
Percent Purity 98%
Chemical Name or Material 4-n-Butoxyphenyl isocyanate
Show More Show Less
Hazard Category H302+H332-H315-H317-H319-H335
Hazard Statement GHS H Statement
H334-H302-H332-H335-H315-H319
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Harmful if swallowed.
Harmful if inhaled.
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement P261-P264b-P270-P271-P272-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P333+P313-P362-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: ISOCYANATES, TOXIC, N.O.S.
TSCA No
Recommended Storage Keep cold

RUO – Research Use Only