missing translation for 'onlineSavingsMsg'
Learn More

4-n-Butoxybenzoyl chloride, 98%

CAS: 33863-86-4 | C11H13ClO2 | 212.673 g/mol

$718.48 - $718.48

Chemical Identifiers

CAS 33863-86-4
Molecular Formula C11H13ClO2
Molecular Weight (g/mol) 212.673
MDL Number MFCD00000689
InChI Key KMGCTFHTBKBITO-UHFFFAOYSA-N
PubChem CID 118561
IUPAC Name 4-butoxybenzoyl chloride
SMILES CCCCOC1=CC=C(C=C1)C(=O)Cl
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAA1165814
View Documents
Thermo Scientific Chemicals
A1165814
25 g N/A
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

33863-86-4
212.673
KMGCTFHTBKBITO-UHFFFAOYSA-N
4-butoxybenzoyl chloride
C11H13ClO2
MFCD00000689
118561
CCCCOC1=CC=C(C=C1)C(=O)Cl

Specifications

33863-86-4
161°C to 162°C (9 mmHg)
C11H13ClO2
MFCD00000689
UN3265
Moisture sensitive
KMGCTFHTBKBITO-UHFFFAOYSA-N
4-butoxybenzoyl chloride
118561
98%
1.121
>110°C (230°F)
1.5495
25 g
957203
4-n-butoxybenzoyl chloride, 4-butoxy-benzoyl chloride, benzoyl chloride, 4-butoxy, p-butoxybenzoyl chloride, 4-n-butoxybenzoylchloride, 4-butyloxy benzoyl chloride, benzoyl chloride,4-butoxy, 4-n-butoxy benzoyl chloride, 4-n-butyloxy benzoyl chloride
CCCCOC1=CC=C(C=C1)C(=O)Cl
212.673
212.68
4-n-Butoxybenzoyl chloride
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.

P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c

H314-H335

DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.

EINECSNumber : 251-703-6

TSCA : No

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only