missing translation for 'onlineSavingsMsg'
Learn More

4-N-BOC-Aminopiperidine, 98%

CAS: 73874-95-0 | C10H21N2O2 | 201.29 g/mol

$269.91 - $269.91

Chemical Identifiers

CAS 73874-95-0
Molecular Formula C10H21N2O2
Molecular Weight (g/mol) 201.29
MDL Number MFCD00798171
InChI Key CKXZPVPIDOJLLM-UHFFFAOYSA-O
Synonym 4-boc-aminopiperidine, tert-butyl piperidin-4-ylcarbamate, 4-n-boc-aminopiperidine, 4-boc-amino piperidine, 4-n-boc-amino piperidine, tert-butyl n-piperidin-4-yl carbamate, 4-tert-butoxycarbonylamino piperidine, 4-n-boc-amino-piperidine, 4-boc-amino-piperidine, 4-n-boc amino piperidine
PubChem CID 723833
IUPAC Name tert-butyl N-piperidin-4-ylcarbamate
SMILES CC(C)(C)OC(=O)NC1CC[NH2+]CC1
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC367620050
SDS View Documents
Thermo Scientific Chemicals
367620050
5 g Glass Bottle
Each for $269.91
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

73874-95-0
201.29
CKXZPVPIDOJLLM-UHFFFAOYSA-O
723833
CC(C)(C)OC(=O)NC1CC[NH2+]CC1
C10H21N2O2
MFCD00798171
4-boc-aminopiperidine, tert-butyl piperidin-4-ylcarbamate, 4-n-boc-aminopiperidine, 4-boc-amino piperidine, 4-n-boc-amino piperidine, tert-butyl n-piperidin-4-yl carbamate, 4-tert-butoxycarbonylamino piperidine, 4-n-boc-amino-piperidine, 4-boc-amino-piperidine, 4-n-boc amino piperidine
tert-butyl N-piperidin-4-ylcarbamate

Specifications

162.0°C to 166.0°C
97.5
White
98%
Glass Bottle
4-boc-aminopiperidine, tert-butyl piperidin-4-ylcarbamate, 4-n-boc-aminopiperidine, 4-boc-amino piperidine, 4-n-boc-amino piperidine, tert-butyl n-piperidin-4-yl carbamate, 4-tert-butoxycarbonylamino piperidine, 4-n-boc-amino-piperidine, 4-boc-amino-piperidine, 4-n-boc amino piperidine
CC(C)(C)OC(=O)NC1CC[NH2+]CC1
201.29
723833
98%
4-N-BOC-Aminopiperidine, 98%
73874-95-0
100.0
Authentic
C10H21N2O2
MFCD00798171
CKXZPVPIDOJLLM-UHFFFAOYSA-O
tert-butyl N-piperidin-4-ylcarbamate
5 g
200.28
Crystalline Powder
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes serious eye damage.
Causes severe skin burns and eye damage.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only