missing translation for 'onlineSavingsMsg'
Learn More

4-(N-BOC-amino)phenylboronic acid, 97%, Thermo Scientific™

Chemical Identifiers

CAS 380430-49-9
Molecular Formula C11H16BNO4
Molecular Weight (g/mol) 237.08
MDL Number MFCD02093054
InChI Key UBVOLHQIEQVXGM-UHFFFAOYSA-N
Synonym 4-boc-aminophenyl boronic acid, 4-n-boc-amino phenylboronic acid, 4-tert-butoxycarbonyl amino phenyl boronic acid, 4-tert-butoxycarbonylamino phenylboronic acid, 4-tert-butoxycarbonyl aminophenylboronic acid, 4-boc-amino benzeneboronic acid, 4-aminobenzeneboronic acid, n-boc protected, 4-tert-butoxycarbonyl amino phenylboronic acid, 4-2-methylpropan-2-yl oxycarbonylamino phenyl boronic acid, 4-tert-butoxycarbonylamino phenyl boronic acid
PubChem CID 3613184
IUPAC Name [4-[(2-methylpropan-2-yl)oxycarbonylamino]phenyl]boronic acid
SMILES B(C1=CC=C(C=C1)NC(=O)OC(C)(C)C)(O)O
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC397030010
SDS View Documents
Thermo Scientific Chemicals
397030010
1g Glass bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

380430-49-9
237.08
UBVOLHQIEQVXGM-UHFFFAOYSA-N
3613184
B(C1=CC=C(C=C1)NC(=O)OC(C)(C)C)(O)O
C11H16BNO4
MFCD02093054
4-boc-aminophenyl boronic acid, 4-n-boc-amino phenylboronic acid, 4-tert-butoxycarbonyl amino phenyl boronic acid, 4-tert-butoxycarbonylamino phenylboronic acid, 4-tert-butoxycarbonyl aminophenylboronic acid, 4-boc-amino benzeneboronic acid, 4-aminobenzeneboronic acid, n-boc protected, 4-tert-butoxycarbonyl amino phenylboronic acid, 4-2-methylpropan-2-yl oxycarbonylamino phenyl boronic acid, 4-tert-butoxycarbonylamino phenyl boronic acid
[4-[(2-methylpropan-2-yl)oxycarbonylamino]phenyl]boronic acid

Specifications

380430-49-9
100.0
96% min. (HPLC)
C11H16BNO4
1g
UBVOLHQIEQVXGM-UHFFFAOYSA-N
[4-[(2-methylpropan-2-yl)oxycarbonylamino]phenyl]boronic acid
3613184
97%
96.0
Authentic
Glass bottle
MFCD02093054
4-boc-aminophenyl boronic acid, 4-n-boc-amino phenylboronic acid, 4-tert-butoxycarbonyl amino phenyl boronic acid, 4-tert-butoxycarbonylamino phenylboronic acid, 4-tert-butoxycarbonyl aminophenylboronic acid, 4-boc-amino benzeneboronic acid, 4-aminobenzeneboronic acid, n-boc protected, 4-tert-butoxycarbonyl amino phenylboronic acid, 4-2-methylpropan-2-yl oxycarbonylamino phenyl boronic acid, 4-tert-butoxycarbonylamino phenyl boronic acid
B(C1=CC=C(C=C1)NC(=O)OC(C)(C)C)(O)O
237.08
237.08
4-(N-BOC-amino)phenylboronic acid
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
Harmful if swallowed.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

SDS
Documents

Documents

RUO – Research Use Only