missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Morpholinophenylboronic acid, 97%, Thermo Scientific™
Specifications
Specifications
| Assay Percent Range | 97% |
| Packaging | Amber Glass Bottle |
| Quantity | 250 mg |
| Formula Weight | 207.04 |
| Percent Purity | 97% |
| Chemical Name or Material | 4-Morpholinophenylboronic acid |