missing translation for 'onlineSavingsMsg'
Learn More

4-Morpholinoacetophenone, 99%, Thermo Scientific™

Chemical Identifiers

CAS 39910-98-0
Molecular Formula C12H15NO2
Molecular Weight (g/mol) 205.26
MDL Number MFCD00006168
InChI Key AKQWEDMTPCAESO-UHFFFAOYSA-N
Synonym 4'-morpholinoacetophenone, 1-4-morpholinophenyl ethanone, 4-morpholinoacetophenone, acetophenone, 4'-morpholino, ethanone, 1-4-4-morpholinyl phenyl, p-morpholinoacetophenone, 1-4-morpholin-4-yl phenyl ethan-1-one, p-morpholino acetophenone, 4;-morpholinoacetophenone, 1-4-morpholin-4-ylphenyl ethanone
PubChem CID 38362
IUPAC Name 1-(4-morpholin-4-ylphenyl)ethanone
SMILES CC(=O)C1=CC=C(C=C1)N2CCOCC2
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC152900050
SDS View Documents
Thermo Scientific Chemicals
152900050
5g Glass bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC152900500
SDS View Documents
Thermo Scientific Chemicals
152900500
50g Glass bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

39910-98-0
205.26
AKQWEDMTPCAESO-UHFFFAOYSA-N
38362
CC(=O)C1=CC=C(C=C1)N2CCOCC2
C12H15NO2
MFCD00006168
4'-morpholinoacetophenone, 1-4-morpholinophenyl ethanone, 4-morpholinoacetophenone, acetophenone, 4'-morpholino, ethanone, 1-4-4-morpholinyl phenyl, p-morpholinoacetophenone, 1-4-morpholin-4-yl phenyl ethan-1-one, p-morpholino acetophenone, 4;-morpholinoacetophenone, 1-4-morpholin-4-ylphenyl ethanone
1-(4-morpholin-4-ylphenyl)ethanone

Specifications

39910-98-0
100.0
98.5% min. (GC)
C12H15NO2
5g
AKQWEDMTPCAESO-UHFFFAOYSA-N
1-(4-morpholin-4-ylphenyl)ethanone
38362
99%
98.5
Authentic
Glass bottle
MFCD00006168
4'-morpholinoacetophenone, 1-4-morpholinophenyl ethanone, 4-morpholinoacetophenone, acetophenone, 4'-morpholino, ethanone, 1-4-4-morpholinyl phenyl, p-morpholinoacetophenone, 1-4-morpholin-4-yl phenyl ethan-1-one, p-morpholino acetophenone, 4;-morpholinoacetophenone, 1-4-morpholin-4-ylphenyl ethanone
CC(=O)C1=CC=C(C=C1)N2CCOCC2
205.26
205.26
4-Morpholinoacetophenone, 99%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
If eye irritation persists: Get medical advice/attention.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Call a POISON CENTER

GHS Signal Word: Warning

EINECSNumber : 254-698-9

RTECSNumber : AM9486200

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only