missing translation for 'onlineSavingsMsg'
Learn More

4-Morpholinepropionitrile, 98+%

CAS: 4542-47-6 | C7H12N2O | 140.186 g/mol

$162.25 - $162.25

Chemical Identifiers

CAS 4542-47-6
Molecular Formula C7H12N2O
Molecular Weight (g/mol) 140.186
MDL Number MFCD00006178
InChI Key WXVKGHVDWWXBJX-UHFFFAOYSA-N
Synonym 3-morpholinopropionitrile, 3-morpholin-4-yl propanenitrile, 3-4-morpholino propionitrile, 4-morpholinepropanenitrile, cyanoethylmorpholine, 4-morpholinepropionitrile, beta-morpholinopropionitrile, n-2-cyanoethyl morpholine, propionitrile, 3-morpholino, 3-morpholinopropanenitrile
PubChem CID 78298
IUPAC Name 3-morpholin-4-ylpropanenitrile
SMILES C1COCCN1CCC#N
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAL1166414
View Documents
Thermo Scientific Chemicals
L1166414
25 g N/A
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

4542-47-6
140.186
WXVKGHVDWWXBJX-UHFFFAOYSA-N
78298
C1COCCN1CCC#N
C7H12N2O
MFCD00006178
3-morpholinopropionitrile, 3-morpholin-4-yl propanenitrile, 3-4-morpholino propionitrile, 4-morpholinepropanenitrile, cyanoethylmorpholine, 4-morpholinepropionitrile, beta-morpholinopropionitrile, n-2-cyanoethyl morpholine, propionitrile, 3-morpholino, 3-morpholinopropanenitrile
3-morpholin-4-ylpropanenitrile

Specifications

4542-47-6
1.037
>110°C (230°F)
1.472
25 g
110425
WXVKGHVDWWXBJX-UHFFFAOYSA-N
3-morpholin-4-ylpropanenitrile
78298
≥98%
20°C to 21°C
106°C to 109°C (7 mmHg)
C7H12N2O
MFCD00006178
UN3276
3-morpholinopropionitrile, 3-morpholin-4-yl propanenitrile, 3-4-morpholino propionitrile, 4-morpholinepropanenitrile, cyanoethylmorpholine, 4-morpholinepropionitrile, beta-morpholinopropionitrile, n-2-cyanoethyl morpholine, propionitrile, 3-morpholino, 3-morpholinopropanenitrile
C1COCCN1CCC#N
140.186
140.19
4-Morpholinepropionitrile
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H331-H302-H312-H315-H319
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

H302+H312+H332-H315-H319

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITRILES, LIQUID, TOXIC, N.O.S.

EINECSNumber : 224-894-9

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only