missing translation for 'onlineSavingsMsg'
Learn More

4-Morpholinepropionitrile, 98+%

Catalog No. AAL1166414
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1166414 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAL1166414 Supplier Thermo Scientific Chemicals Supplier No. L1166414

CAS: 4542-47-6 | C7H12N2O | 140.186 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 4542-47-6
Molecular Formula C7H12N2O
Molecular Weight (g/mol) 140.186
MDL Number MFCD00006178
InChI Key WXVKGHVDWWXBJX-UHFFFAOYSA-N
Synonym 3-morpholinopropionitrile, 3-morpholin-4-yl propanenitrile, 3-4-morpholino propionitrile, 4-morpholinepropanenitrile, cyanoethylmorpholine, 4-morpholinepropionitrile, beta-morpholinopropionitrile, n-2-cyanoethyl morpholine, propionitrile, 3-morpholino, 3-morpholinopropanenitrile
PubChem CID 78298
IUPAC Name 3-morpholin-4-ylpropanenitrile
SMILES C1COCCN1CCC#N

Specifications

Melting Point 20°C to 21°C
Density 1.037
Boiling Point 106°C to 109°C (7 mmHg)
Flash Point >110°C (230°F)
Refractive Index 1.472
Quantity 25 g
UN Number UN3276
Beilstein 110425
Formula Weight 140.19
Percent Purity ≥98%
Chemical Name or Material 4-Morpholinepropionitrile
Show More Show Less
Hazard Category H302+H312+H332-H315-H319
Hazard Statement GHS H Statement
H331-H302-H312-H315-H319
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITRILES, LIQUID, TOXIC, N.O.S.
EINECSNumber 224-894-9
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only