missing translation for 'onlineSavingsMsg'
Learn More

4-Methylvaleric acid, 99%

Catalog No. p-169127
Change view
Click to view available options
Quantity:
5 mL
25 mL
Packaging:
Glass bottle
Catalog No. Quantity Packaging
AC255510050 5 mL Glass bottle
AC255510250 25 mL Glass bottle
2 options

Catalog No. AC255510050

Supplier: Thermo Scientific Chemicals 255510050

Only null left

CAS: 646-07-1 | C6H12O2 | 116.15 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 646-07-1
Molecular Formula C6H12O2
Molecular Weight (g/mol) 116.15
MDL Number MFCD00002803
InChI Key FGKJLKRYENPLQH-UHFFFAOYSA-N
Synonym 4-methylvaleric acid, isocaproic acid, isohexanoic acid, pentanoic acid, 4-methyl, isobutylacetic acid, 4-methyl-pentanoic acid, valeric acid, 4-methyl, 4-methyl valeric acid, isocaproate, 4,4-dimethylbutanoic acid
PubChem CID 12587
ChEBI CHEBI:74903
IUPAC Name 4-methylpentanoic acid
SMILES CC(C)CCC(=O)O

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point -35.0°C
Color Colorless to Yellow
Density 0.9220g/mL
Boiling Point 200.0°C
Flash Point 97°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.4136 to 1.4156
Linear Formula (CH3)2CHCH2CH2COOH
Quantity 5 mL
Specific Gravity 0.922
Solubility Information Solubility in water: insoluble
Formula Weight 116.15
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material 4-Methylvaleric acid, 99%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Toxic in contact with skin.
Causes severe skin burns and eye damage.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with wat
EINECSNumber 211-464-0
RTECSNumber NR2975000
TSCA TSCA

RUO – Research Use Only