missing translation for 'onlineSavingsMsg'
Learn More

4-Methylvaleric acid, 99%

CAS: 646-07-1 | C6H12O2 | 116.16 g/mol

Supplier:  Thermo Scientific Chemicals A1540514

 View more versions of this product

Catalog No. AAA1540514


Only null left
Explore available promotions

Description

Description

4-Methylvaleric acid has been used in asymmetric synthesis of (4S)-4-isopropyl-3-[(2?S)-2?,4?-dimethyl-valeryl)]-2-oxazolidinone. It is a versatile building block used in the synthesis of various pharmaceutical and biologically active compounds, including inhibitors and antagonists. It is involved in the synthesis of novel series of high affinity, functionally potent antagonists of the CCR1 receptor.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Methylvaleric acid has been used in asymmetric synthesis of (4S)-4-isopropyl-3-[(2′S)-2′,4′-dimethyl-valeryl)]-2-oxazolidinone. It is a versatile building block used in the synthesis of various pharmaceutical and biologically active compounds, including inhibitors and antagonists. It is involved in the synthesis of novel series of high affinity, functionally potent antagonists of the CCR1 receptor.

Solubility
Soluble in alcohol. Slightly soluble in water.

Notes
Store in cool, dry conditions in a well sealed container. Store away from oxidizing agents, bases.
Specifications

Specifications

4-Methylvaleric acid
-35°C
199°C to 201°C
Unpleasant
1.4146
25 g
1741912
Soluble in alcohol. Slightly soluble in water.
CC(C)CCC(=O)O
116.16
CHEBI:74903
99%
646-07-1
0.923
97°C (206°F)
C6H12O2
MFCD00002803
UN2922
4-methylvaleric acid, isocaproic acid, isohexanoic acid, pentanoic acid, 4-methyl, isobutylacetic acid, 4-methyl-pentanoic acid, valeric acid, 4-methyl, 4-methyl valeric acid, isocaproate, 4,4-dimethylbutanoic acid
FGKJLKRYENPLQH-UHFFFAOYSA-N
4-methylpentanoic acid
12587
116.16

Chemical Identifiers

646-07-1
116.16
FGKJLKRYENPLQH-UHFFFAOYSA-N
12587
4-methylpentanoic acid
C6H12O2
MFCD00002803
4-methylvaleric acid, isocaproic acid, isohexanoic acid, pentanoic acid, 4-methyl, isobutylacetic acid, 4-methyl-pentanoic acid, valeric acid, 4-methyl, 4-methyl valeric acid, isocaproate, 4,4-dimethylbutanoic acid
CHEBI:74903
CC(C)CCC(=O)O
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H311-H314-H318
Toxic in contact with skin.
Causes severe skin burns and eye damage.
Causes serious eye damage.

P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c

H311-H314

DOTInformation : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE LIQUIDS, TOXIC, N.O.S.

EINECSNumber : 211-464-0

RTECSNumber : NR2975000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only