missing translation for 'onlineSavingsMsg'
Learn More

4-Methylumbelliferyl beta-D-Glucuronide Hydrate 98.0+%, TCI America™

Catalog No. M3026100MG
Change view
Click to view available options
Quantity:
100 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M3026100MG 100 mg
M30261G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M3026100MG Supplier TCI America Supplier No. M3026100MG

Chemical Identifiers

CAS 881005-91-0
Molecular Formula C16H18O10
Molecular Weight (g/mol) 370.31
MDL Number MFCD09039280
InChI Key URVSQZMOFUEQAW-YYHOVTOASA-N
PubChem CID 16218413
IUPAC Name (2S,3S,4S,5R,6S)-3,4,5-trihydroxy-6-(4-methyl-2-oxochromen-7-yl)oxyoxane-2-carboxylic acid;hydrate
SMILES CC1=CC(=O)OC2=C1C=CC(=C2)OC3C(C(C(C(O3)C(=O)O)O)O)O.O

Specifications

Color White
Quantity 100 mg
Formula Weight 352.30
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material 4-Methylumbelliferyl beta-D-Glucuronide Hydrate