missing translation for 'onlineSavingsMsg'
Learn More

4-Methylumbelliferyl-beta-D-galactopyranoside, 97+%

CAS: 6160-78-7 | C16H18O8 | 338.31 g/mol

$99.90 - $282.45

Chemical Identifiers

CAS 6160-78-7
Molecular Formula C16H18O8
Molecular Weight (g/mol) 338.31
MDL Number MFCD00036773,MFCD00063694
InChI Key YUDPTGPSBJVHCN-DZQJYWQESA-N
Synonym 4-methylumbelliferyl-beta-d-galactopyranoside, 4-methylumbelliferyl-galactopyranoside, 4-methylumbelliferyl beta-d-galactoside, 4-methylumbelliferyl b-d-galactoside, muga, 4-methylumbelliferyl beta-galactoside, 7-beta-d-galactopyranosyloxy-4-methyl-2h-1-benzopyran-2-one, 4-methylumbelliferyl b-d-galactopyranoside, 4-methyl-7-2s,3r,4s,5r,6r-3,4,5-trihydroxy-6-hydroxymethyl tetrahydropyran-2-yl oxy-chromen-2-one, 4-methyl-7-2s,3r,4s,5r,6r-3,4,5-trihydroxy-6-hydroxymethyl oxan-2-yl oxy chromen-2-one
PubChem CID 93577
IUPAC Name 4-methyl-7-[(2S,3R,4S,5R,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl]oxychromen-2-one
SMILES CC1=CC(=O)OC2=CC(O[C@@H]3O[C@H](CO)[C@H](O)[C@H](O)[C@H]3O)=CC=C12
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAB2107501
View Documents
Thermo Scientific Chemicals
B2107501
0.25 g
Each for $99.90
Only null left
 
AAB2107503
View Documents
Thermo Scientific Chemicals
B2107503
1 g
Each for $282.45
Only null left
 
Description

Description

It is a substrate used for fluorometric assay for β-galactosidase. It is used to detect E-coli n the fluorogenic medium.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is a substrate used for fluorometric assay for β-galactosidase. It is used to detect E-coli n the fluorogenic medium.

Solubility
Soluble in water.

Notes
Moisture sensitive. Store at -20°C. Keep away from oxidizing agents.
Specifications

Chemical Identifiers

6160-78-7
338.31
YUDPTGPSBJVHCN-DZQJYWQESA-N
93577
CC1=CC(=O)OC2=CC(O[C@@H]3O[C@H](CO)[C@H](O)[C@H](O)[C@H]3O)=CC=C12
C16H18O8
MFCD00036773,MFCD00063694
4-methylumbelliferyl-beta-d-galactopyranoside, 4-methylumbelliferyl-galactopyranoside, 4-methylumbelliferyl beta-d-galactoside, 4-methylumbelliferyl b-d-galactoside, muga, 4-methylumbelliferyl beta-galactoside, 7-beta-d-galactopyranosyloxy-4-methyl-2h-1-benzopyran-2-one, 4-methylumbelliferyl b-d-galactopyranoside, 4-methyl-7-2s,3r,4s,5r,6r-3,4,5-trihydroxy-6-hydroxymethyl tetrahydropyran-2-yl oxy-chromen-2-one, 4-methyl-7-2s,3r,4s,5r,6r-3,4,5-trihydroxy-6-hydroxymethyl oxan-2-yl oxy chromen-2-one
4-methyl-7-[(2S,3R,4S,5R,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl]oxychromen-2-one

Specifications

6160-78-7
C16H18O8
0.25 g
Light sensitive
Soluble in water.
−64° (c=1 in Pyridine, 24h)
4-methyl-7-[(2S,3R,4S,5R,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl]oxychromen-2-one
93577
98%
∼260°C (decomposition)
MFCD00036773,MFCD00063694
94673
4-methylumbelliferyl-beta-d-galactopyranoside, 4-methylumbelliferyl-galactopyranoside, 4-methylumbelliferyl beta-d-galactoside, 4-methylumbelliferyl b-d-galactoside, muga, 4-methylumbelliferyl beta-galactoside, 7-beta-d-galactopyranosyloxy-4-methyl-2h-1-benzopyran-2-one, 4-methylumbelliferyl b-d-galactopyranoside, 4-methyl-7-2s,3r,4s,5r,6r-3,4,5-trihydroxy-6-hydroxymethyl tetrahydropyran-2-yl oxy-chromen-2-one, 4-methyl-7-2s,3r,4s,5r,6r-3,4,5-trihydroxy-6-hydroxymethyl oxan-2-yl oxy chromen-2-one
YUDPTGPSBJVHCN-DZQJYWQESA-N
CC1=CC(=O)OC2=CC(O[C@@H]3O[C@H](CO)[C@H](O)[C@H](O)[C@H]3O)=CC=C12
338.31
338.32
4-Methylumbelliferyl-beta-D-galactopyranoside
Videos
Safety and Handling

Safety and Handling

EINECSNumber : 228-185-5

TSCA : No

Recommended Storage : Store at -20°C

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only