missing translation for 'onlineSavingsMsg'
Learn More

4-Methylumbelliferyl-beta-D-galactopyranoside, 97+%

Catalog No. AAB2107501
Change view
Click to view available options
Quantity:
0.25 g
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2107501 0.25 g
AAB2107503 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2107501 Supplier Thermo Scientific Chemicals Supplier No. B2107501
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 6160-78-7 | C16H18O8 | 338.31 g/mol

It is a substrate used for fluorometric assay for β-galactosidase. It is used to detect E-coli n the fluorogenic medium.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is a substrate used for fluorometric assay for β-galactosidase. It is used to detect E-coli n the fluorogenic medium.

Solubility
Soluble in water.

Notes
Moisture sensitive. Store at -20°C. Keep away from oxidizing agents.

Chemical Identifiers

CAS 6160-78-7
Molecular Formula C16H18O8
Molecular Weight (g/mol) 338.31
MDL Number MFCD00036773,MFCD00063694
InChI Key YUDPTGPSBJVHCN-DZQJYWQESA-N
Synonym 4-methylumbelliferyl-beta-d-galactopyranoside, 4-methylumbelliferyl-galactopyranoside, 4-methylumbelliferyl beta-d-galactoside, 4-methylumbelliferyl b-d-galactoside, muga, 4-methylumbelliferyl beta-galactoside, 7-beta-d-galactopyranosyloxy-4-methyl-2h-1-benzopyran-2-one, 4-methylumbelliferyl b-d-galactopyranoside, 4-methyl-7-2s,3r,4s,5r,6r-3,4,5-trihydroxy-6-hydroxymethyl tetrahydropyran-2-yl oxy-chromen-2-one, 4-methyl-7-2s,3r,4s,5r,6r-3,4,5-trihydroxy-6-hydroxymethyl oxan-2-yl oxy chromen-2-one
PubChem CID 93577
IUPAC Name 4-methyl-7-[(2S,3R,4S,5R,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl]oxychromen-2-one
SMILES CC1=CC(=O)OC2=CC(O[C@@H]3O[C@H](CO)[C@H](O)[C@H](O)[C@H]3O)=CC=C12

Specifications

Melting Point ∼260°C (decomposition)
Quantity 0.25 g
Beilstein 94673
Sensitivity Light sensitive
Solubility Information Soluble in water.
Optical Rotation −64° (c=1 in Pyridine, 24h)
Formula Weight 338.32
Percent Purity 98%
Chemical Name or Material 4-Methylumbelliferyl-beta-D-galactopyranoside
EINECSNumber 228-185-5
TSCA No
Recommended Storage Store at -20°C

RUO – Research Use Only