missing translation for 'onlineSavingsMsg'
Learn More

4-Methylthiazole-5-carboxaldehyde 98.0+%, TCI America™

Catalog No. M18125G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M18125G 5 g
M18121G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M18125G Supplier TCI America Supplier No. M18125G

Chemical Identifiers

CAS 82294-70-0
Molecular Formula C5H5NOS
Molecular Weight (g/mol) 127.161
MDL Number MFCD07368277
InChI Key JJVIEMFQPALZOZ-UHFFFAOYSA-N
Synonym 4-methylthiazole-5-carbaldehyde, 4-methyl-5-thiazolecarboxaldehyde, 4-methylthiazole-5-carboxaldehyde, 4-methyl-5-formylthiazole, 4-methyl-5-thiazolecarbaldehyde, 5-formyl-4-methylthiazole, 4-methylthiazole-5-aldehyde, 4-methyl-5-thiazolyl aldehyde, 5-thiazolecarboxaldehyde, 4-methyl, 4-methyl-5-thiazole carboxaldehyde
PubChem CID 581339
IUPAC Name 4-methyl-1,3-thiazole-5-carbaldehyde
SMILES CC1=C(SC=N1)C=O

Specifications

Melting Point 76°C
Boiling Point 118°C
Quantity 5 g
Formula Weight 127.16
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4-Methylthiazole-5-carboxaldehyde