missing translation for 'onlineSavingsMsg'
Learn More

4'-(Methylsulfonyl)acetophenone 98.0+%, TCI America™

Catalog No. p-7132803
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
25 g
250 g
Catalog No. Quantity
M167825G 25 g
M1678250G 250 g
2 options

Catalog No. M167825G

Supplier: TCI America M167825G

Only null left

Chemical Identifiers

CAS 10297-73-1
Molecular Formula C9H10O3S
Molecular Weight (g/mol) 198.236
InChI Key KAVZYDHKJNABPC-UHFFFAOYSA-N
Synonym 4'-methylsulfonyl acetophenone, 1-4-methylsulfonyl phenyl ethanone, 4-methylsulfonylacetophenone, 4-methylsulfonyl acetophenone, 4-acetylphenyl methyl sulphone, 1-4-methylsulfonylphenyl ethanone, 4-methylsulphonylacetophenone, 1-4-methanesulfonyl-phenyl-ethanone, p-methylsulfonylacetophenone, 4-methanesulfonylacetophenone
PubChem CID 82529
IUPAC Name 1-(4-methylsulfonylphenyl)ethanone
SMILES CC(=O)C1=CC=C(C=C1)S(=O)(=O)C

Specifications

Melting Point 129°C
Color White-Yellow
Quantity 25 g
Formula Weight 198.24
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4′-(Methylsulfonyl)acetophenone