missing translation for 'onlineSavingsMsg'
Learn More

4'-(Methylsulfonyl)acetophenone 98.0+%, TCI America™

Catalog No. M167825G
Change view
Click to view available options
Quantity:
25 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M167825G 25 g
M1678250G 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M167825G Supplier TCI America Supplier No. M167825G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 10297-73-1
Molecular Formula C9H10O3S
Molecular Weight (g/mol) 198.236
InChI Key KAVZYDHKJNABPC-UHFFFAOYSA-N
Synonym 4'-methylsulfonyl acetophenone, 1-4-methylsulfonyl phenyl ethanone, 4-methylsulfonylacetophenone, 4-methylsulfonyl acetophenone, 4-acetylphenyl methyl sulphone, 1-4-methylsulfonylphenyl ethanone, 4-methylsulphonylacetophenone, 1-4-methanesulfonyl-phenyl-ethanone, p-methylsulfonylacetophenone, 4-methanesulfonylacetophenone
PubChem CID 82529
IUPAC Name 1-(4-methylsulfonylphenyl)ethanone
SMILES CC(=O)C1=CC=C(C=C1)S(=O)(=O)C

Specifications

Melting Point 129°C
Color White-Yellow
Quantity 25 g
Formula Weight 198.24
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4′-(Methylsulfonyl)acetophenone