missing translation for 'onlineSavingsMsg'
Learn More

4-Methylstilbene 98.0+%, TCI America™

Catalog No. M14925G
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M14925G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M14925G Supplier TCI America Supplier No. M14925G
Only null left

Chemical Identifiers

CAS 4714-21-0
Molecular Formula C15H14
Molecular Weight (g/mol) 194.277
MDL Number MFCD00452726
InChI Key MDRVHDXASYPUCB-VAWYXSNFSA-N
PubChem CID 5314921
IUPAC Name 1-methyl-4-[(E)-2-phenylethenyl]benzene
SMILES CC1=CC=C(C=C1)C=CC2=CC=CC=C2

Specifications

Melting Point 120°C
Color White
Quantity 5 g
Formula Weight 194.28
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4-Methylstilbene