missing translation for 'onlineSavingsMsg'
Learn More

4-Methylsalicylaldehyde 98.0+%, TCI America™

Catalog No. p-7133217
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M24315G 5 g
M243125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M24315G Supplier TCI America Supplier No. M24315G

Chemical Identifiers

CAS 698-27-1
Molecular Formula C8H8O2
Molecular Weight (g/mol) 136.15
MDL Number MFCD00799550
InChI Key JODRRPJMQDFCBJ-UHFFFAOYSA-N
Synonym 4-methylsalicylaldehyde, 2,4-cresotaldehyde, benzaldehyde, 2-hydroxy-4-methyl, 4-methyl-2-hydroxybenzaldehyde, 2-hydroxy-4-methyl-benzaldehyde, 2-formyl-5-methylphenol, 4-methylsalicyclic aldehyde, unii-9n0s26g61j, 4-methyl salicylaldehyde, homosalicylaldehyde, m
PubChem CID 61200
IUPAC Name 2-hydroxy-4-methylbenzaldehyde
SMILES CC1=CC=C(C=O)C(O)=C1

Specifications

Melting Point 62°C
Color White-Yellow
Boiling Point 223°C
Quantity 5 g
Formula Weight 136.15
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 4-Methylsalicylaldehyde