missing translation for 'onlineSavingsMsg'
Learn More

4-Methylquinoline, 99%

CAS: 491-35-0 | C10H9N | 143.19 g/mol

$95.49 - $219.83

Chemical Identifiers

CAS 491-35-0
Molecular Formula C10H9N
Molecular Weight (g/mol) 143.19
MDL Number MFCD00006784
InChI Key MUDSDYNRBDKLGK-UHFFFAOYSA-N
Synonym lepidine, quinoline, 4-methyl, cincholepidine, lepidin, p-methylquinoline, 4-lepidine, gamma-methylquinoline, 4-methyl-quinoline, 4-methyl quinoline, ccris 2894
PubChem CID 10285
ChEBI CHEBI:48983
IUPAC Name 4-methylquinoline
SMILES CC1=CC=NC2=CC=CC=C12
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC169770050
SDS View Documents
Thermo Scientific Chemicals
169770050
5 g Glass bottle
Each for $95.49
Only null left
 
AC169770250
SDS View Documents
Thermo Scientific Chemicals
169770250
25 g Glass bottle
Each for $219.83
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

491-35-0
143.19
MUDSDYNRBDKLGK-UHFFFAOYSA-N
10285
4-methylquinoline
C10H9N
MFCD00006784
lepidine, quinoline, 4-methyl, cincholepidine, lepidin, p-methylquinoline, 4-lepidine, gamma-methylquinoline, 4-methyl-quinoline, 4-methyl quinoline, ccris 2894
CHEBI:48983
CC1=CC=NC2=CC=CC=C12

Specifications

491-35-0
1.0800g/mL
>110°C
98.5% min. (GC)
C10H9N
MFCD00006784
20, 395
15, 5496
Solubility in water: slightly soluble. Other solubilities: miscible with alcohol,benzene and ether
CC1=CC=NC2=CC=CC=C12
143.19
CHEBI:48983
99%
4-Methylquinoline, 99%
9.0°C to 10.0°C
261.0°C to 263.0°C
Authentic
Glass bottle
1.6180 to 1.6200
5 g
1.08
lepidine, quinoline, 4-methyl, cincholepidine, lepidin, p-methylquinoline, 4-lepidine, gamma-methylquinoline, 4-methyl-quinoline, 4-methyl quinoline, ccris 2894
MUDSDYNRBDKLGK-UHFFFAOYSA-N
4-methylquinoline
10285
143.19
Liquid
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
May cause respiratory irritation.
Causes serious eye irritation.
Suspected of causing genetic defects.
Causes skin irritation.

GHS P Statement:
Use personal protective equipment as required.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 207-734-2

RTECSNumber : OH0316000

TSCA : TSCA

SDS
Documents

Documents

RUO – Research Use Only