missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Methylpyridine 98.0+%, TCI America™
Specifications
Specifications
| Melting Point | 4°C |
| Color | Yellow |
| Boiling Point | 144°C |
| Quantity | 25 mL |
| UN Number | 2313 |
| Formula Weight | 93.13 |
| Percent Purity | ≥98.0% (GC,T) |
| Physical Form | Liquid |
| Chemical Name or Material | 4-Methylpyridine |
Safety and Handling
| EINECSNumber | (5)-0711 |
| RTECSNumber | UT5425000 |
| TSCA | Yes |