missing translation for 'onlineSavingsMsg'
Learn More

4-Methylpyridine-3-carboxylic acid, 97%

Catalog No. AC429420010
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC429420010 1 g Glass bottle
AC429420050 5 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC429420010 Supplier Thermo Scientific Chemicals Supplier No. 429420010

CAS: 3222-50-2 | C7H7NO2 | 137.14 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 3222-50-2
Molecular Formula C7H7NO2
Molecular Weight (g/mol) 137.14
MDL Number MFCD00128869
InChI Key ZKUZSTXNVMIDCY-UHFFFAOYSA-N
Synonym 4-methylnicotinic acid, 4-methylpyridine-3-carboxylicacid, 4-methyl-3-pyridinecarboxylic acid, 3-pyridinecarboxylic acid, 4-methyl, 4-methyl-nicotinic acid, pubchem16270, acmc-1ags3, ksc223o4t, cbi-bb zero/009371, 3-pyridinecarboxylic acid,4-methyl
PubChem CID 229163
IUPAC Name 4-methylpyridine-3-carboxylic acid
SMILES CC1=C(C=NC=C1)C(=O)O

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 214.0°C to 215.0°C
Color Tan to White
Infrared Spectrum Authentic
Assay Percent Range 96% min. (HPLC)
Packaging Glass bottle
Quantity 1 g
Formula Weight 137.14
Percent Purity 97%
Physical Form Crystalline Powder
Chemical Name or Material 4-Methylpyridine-3-carboxylic acid
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

RUO – Research Use Only