missing translation for 'onlineSavingsMsg'
Learn More

4-Methylpyridine-3-boronic acid, 90%

Catalog No. AC430270010
Change view
Click to view available options
Quantity:
1 g
Packaging:
Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC430270010 1 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC430270010 Supplier Thermo Scientific Chemicals Supplier No. 430270010
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 148546-82-1 | C6H8BNO2 | 136.95 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 148546-82-1
Molecular Formula C6H8BNO2
Molecular Weight (g/mol) 136.95
MDL Number MFCD03788244
InChI Key ASXFMIDIRZPCGK-UHFFFAOYSA-N
Synonym 4-methylpyridine-3-boronic acid, 4-methylpyridin-3-yl boronic acid, 4-methylpyridin-3-yl-3-boronic acid, 4-methyl-3-pyridyl boronic acid, 3-borono-4-picoline, boronic acid, 4-methyl-3-pyridinyl, 4-methyl-3-pyridinyl boronic acid, 4-picoline-3-boronic acid, 4-picoline-5-boronic acid, 4-methyl-3-pyridinyl-boronic acid
PubChem CID 2762711
IUPAC Name (4-methylpyridin-3-yl)boronic acid
SMILES B(C1=C(C=CN=C1)C)(O)O

Specifications

CAS Min % 88.0
CAS Max % 100.0
Color Colorless to Yellow
Infrared Spectrum Authentic
Assay Percent Range 88% min. (HPLC)
Packaging Glass bottle
Quantity 1 g
Formula Weight 136.95
Percent Purity 90%
Physical Form Liquid
Chemical Name or Material 4-Methylpyridine-3-boronic acid
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for

RUO – Research Use Only