missing translation for 'onlineSavingsMsg'
Learn More

4-Methylpyrazole 98.0+%, TCI America™

Catalog No. M07745G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M07745G 5 g
M07741G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M07745G Supplier TCI America Supplier No. M07745G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 7554-65-6
Molecular Formula C4H6N2
Molecular Weight (g/mol) 82.106
MDL Number MFCD00005245
InChI Key RIKMMFOAQPJVMX-UHFFFAOYSA-N
Synonym 4-methylpyrazole, fomepizole, antizol, 1h-pyrazole, 4-methyl, fomepizol, 4-methylpyrazol, fomepizolum, fomepizol inn-spanish, fomepizolum inn-latin, fomepizole usan:inn
PubChem CID 3406
ChEBI CHEBI:5141
IUPAC Name 4-methyl-1H-pyrazole
SMILES CC1=CNN=C1

Specifications

Color White-Yellow
Boiling Point 207°C
Quantity 5 g
Formula Weight 82.11
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Lumps
Chemical Name or Material 4-Methylpyrazole
RTECSNumber UQ7350000
TSCA No