missing translation for 'onlineSavingsMsg'
Learn More

4'-Methylpropiophenone 95.0+%, TCI America™

Catalog No. p-7132351
Change view
Click to view available options
Quantity:
25 mL
500 mL
Catalog No. Quantity
M076825ML 25 mL
M0768500ML 500 mL
2 options

Catalog No. M076825ML

Supplier: TCI America M076825ML

Only null left

Chemical Identifiers

CAS 5337-93-9
Molecular Formula C10H12O
Molecular Weight (g/mol) 148.21
MDL Number MFCD00009312
InChI Key PATYHUUYADUHQS-UHFFFAOYSA-N
Synonym 4'-methylpropiophenone, 1-p-tolyl propan-1-one, p-methylpropiophenone, 4-methylpropiophenone, 1-4-methylphenyl propan-1-one, 1-propanone, 1-4-methylphenyl, p-tolyl ethyl ketone, p-methyl propiophenone, 1-p-tolylpropan-1-one, 1-4-methylphenyl-1-propanone
PubChem CID 21429
ChEBI CHEBI:59335
IUPAC Name 1-(4-methylphenyl)propan-1-one
SMILES CCC(=O)C1=CC=C(C)C=C1

Specifications

Color Yellow
Boiling Point 239°C
Quantity 25 mL
Formula Weight 148.21
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material 4′-Methylpropiophenone
EINECSNumber (3)-3865
TSCA No