missing translation for 'onlineSavingsMsg'
Learn More

4'-Methylpropiophenone 95.0+%, TCI America™

Catalog No. M076825ML
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M076825ML 25 mL
M0768500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M076825ML Supplier TCI America Supplier No. M076825ML
Only null left

Chemical Identifiers

CAS 5337-93-9
Molecular Formula C10H12O
Molecular Weight (g/mol) 148.21
MDL Number MFCD00009312
InChI Key PATYHUUYADUHQS-UHFFFAOYSA-N
Synonym 4'-methylpropiophenone, 1-p-tolyl propan-1-one, p-methylpropiophenone, 4-methylpropiophenone, 1-4-methylphenyl propan-1-one, 1-propanone, 1-4-methylphenyl, p-tolyl ethyl ketone, p-methyl propiophenone, 1-p-tolylpropan-1-one, 1-4-methylphenyl-1-propanone
PubChem CID 21429
ChEBI CHEBI:59335
IUPAC Name 1-(4-methylphenyl)propan-1-one
SMILES CCC(=O)C1=CC=C(C)C=C1

Specifications

Color Yellow
Boiling Point 239°C
Quantity 25 mL
Formula Weight 148.21
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material 4′-Methylpropiophenone
EINECSNumber (3)-3865
TSCA No