missing translation for 'onlineSavingsMsg'
Learn More

4-Methylpiperidine, 98+%

Catalog No. AAL02709AP
Change view
Click to view available options
Quantity:
100 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL02709AP 500 mL
AAL02709AE 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL02709AP Supplier Thermo Scientific Chemicals Supplier No. L02709AP
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 626-58-4 | C6H13N | 99.18 g/mol

4-Methylpiperidine acts as a catalyst in the preparation of stilbene derivatives. It is used as an anti-corrosive compound for iron. It is also used in the synthesis of peptides to remove or deprotection of 9-fluorenylmethyloxycarbonyl (Fmoc) groups.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Methylpiperidine acts as a catalyst in the preparation of stilbene derivatives. It is used as an anti-corrosive compound for iron. It is also used in the synthesis of peptides to remove or deprotection of 9-fluorenylmethyloxycarbonyl (Fmoc) groups.

Solubility
Miscible with water and chloroform.

Notes
Store in a cool place. Keep the container tightly closed in a dry and well-ventilated place. Incompatible with acids, acid chlorides, carbon dioxide and strong oxidizing agents.

Chemical Identifiers

CAS 626-58-4
Molecular Formula C6H13N
Molecular Weight (g/mol) 99.18
MDL Number MFCD00006005
InChI Key UZOFELREXGAFOI-UHFFFAOYSA-N
Synonym 4-pipecoline, piperidine, 4-methyl, gamma-pipecoline, 4-methylpiperidin, .gamma.-pipecoline, 4-methyl-piperidine, 4-methylpiperdine, 4-methylpyperidine, 4-methyl piperidine, hexahydro-g-picoline
PubChem CID 69381
IUPAC Name 4-methylpiperidine
SMILES CC1CCNCC1

Specifications

Melting Point 4°C to 5°C
Density 0.838
pH 13
Boiling Point 123°C to 124°C
Flash Point 13°C (55°F)
Odor Amine-like
Refractive Index 1.4458
Quantity 500 mL
UN Number UN2733
Beilstein 1083
Solubility Information Miscible with water and chloroform.
Formula Weight 99.18
Percent Purity ≥98%
Chemical Name or Material 4-Methylpiperidine
Show More Show Less
Hazard Category H225-H302-H311+H331-H314-H335
Hazard Statement GHS H Statement
H225-H314-H318
Highly flammable liquid and vapor.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: AMINES, FLAMMABLE, CORROSIVE, N.O.S.
EINECSNumber 210-954-1
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only