missing translation for 'onlineSavingsMsg'
Learn More

Sigma Aldrich 4-Methylpiperidine

Catalog No. M73206500ML
Encompass
Change view
Click to view available options
Quantity:
500 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M73206500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M73206500ML Supplier Sigma Aldrich Supplier No. M73206500ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • 4-methylpiperidine can be used to synthesize the following bioactive compounds:
  • Dimethyl bis(4-methylpiperidine-dithiocarbamato-s,sα)-tin(iv)
  • (e)-4-(4α-methylpiperidino-n-alkoxy) stilbenes
  • 4-methylpiperidinoalkylthio and 4-methylpiperidinoalkoxy derivatives of (e)-chalconeit can also act as a deprotecting reagent for the removal of fmoc group from amino acids during solid-phase peptide synthesis

Specifications

CAS 626-58-4
Density 0.838 g/mL (at 25°C (literature))
Boiling Point 124°C (lit.)
Molecular Formula C6H13N
Refractive Index n20/D 1.446 (literature)
Linear Formula C6H13N
MDL Number MFCD00006005
Quantity 500 mL
Synonym gamma-Pipecoline; 4-Pipecoline
Molecular Weight (g/mol) 99.17
Percent Purity 96%
Show More Show Less