missing translation for 'onlineSavingsMsg'
Learn More

4-Methylpiperazine-1-ethanol 98.0+%, TCI America™

Catalog No. p-7133257
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M24995G 5 g
M249925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M24995G Supplier TCI America Supplier No. M24995G

Chemical Identifiers

CAS 5464-12-0
Molecular Formula C7H16N2O
Molecular Weight (g/mol) 144.218
MDL Number MFCD03840691
InChI Key QHTUMQYGZQYEOZ-UHFFFAOYSA-N
Synonym 1-2-hydroxyethyl-4-methylpiperazine, 2-4-methylpiperazin-1-yl ethanol, 1-piperazineethanol, 4-methyl, 2-4-methylpiperazin-1-yl ethan-1-ol, 1-n-hydroxyethyl-4-methyl piperazine, 2-4-methyl-piperazin-1-yl-ethanol, 1-2-hydroxyethyl-4-methyl-piperazine, 4-methylpiperazine-1-ethanol, 2-4-methylpiperazinyl ethan-1-ol, 2-4-methyl-1-piperazinyl ethanol
PubChem CID 231184
IUPAC Name 2-(4-methylpiperazin-1-yl)ethanol
SMILES CN1CCN(CC1)CCO

Specifications

Melting Point 41°C
Color White-Yellow
Boiling Point 55°C
Quantity 5 g
Formula Weight 144.22
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 4-Methylpiperazine-1-ethanol
EINECSNumber (5)-5745
TSCA No