missing translation for 'onlineSavingsMsg'
Learn More

4-Methylphthalic Anhydride 98.0+%, TCI America™

Catalog No. p-7132236
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M055925G 25 g
M0559500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M055925G Supplier TCI America Supplier No. M055925G
Only null left

Chemical Identifiers

CAS 19438-61-0
Molecular Formula C9H6O3
Molecular Weight (g/mol) 162.144
MDL Number MFCD00041856
InChI Key ZOXBWJMCXHTKNU-UHFFFAOYSA-N
Synonym 4-methylphthalic anhydride, 5-methylisobenzofuran-1,3-dione, 1,3-isobenzofurandione, 5-methyl, unii-2p2740fpck, phthalic anhydride, 4-methyl, dsstox_cid_7805, 5-methyl-1,3-dihydro-2-benzofuran-1,3-dione, dsstox_rid_78575, dsstox_gsid_27805, 4methylphthalic anhydride
PubChem CID 88069
IUPAC Name 5-methyl-2-benzofuran-1,3-dione
SMILES CC1=CC2=C(C=C1)C(=O)OC2=O

Specifications

Melting Point 94°C
Boiling Point 299°C
Quantity 25 g
Formula Weight 162.14
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 4-Methylphthalic Anhydride
EINECSNumber (3)-3815
TSCA Yes