missing translation for 'onlineSavingsMsg'
Learn More

4-Methylphthalic anhydride, 96%

Catalog No. p-3758461
Change view
Click to view available options
Quantity:
25 g
Packaging:
Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC414790250 25 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC414790250 Supplier Thermo Scientific Chemicals Supplier No. 414790250

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 19438-61-0 | C9H6O3 | 162.14 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 19438-61-0
Molecular Formula C9H6O3
Molecular Weight (g/mol) 162.14
MDL Number MFCD00041856
InChI Key ZOXBWJMCXHTKNU-UHFFFAOYSA-N
Synonym 4-methylphthalic anhydride, 5-methylisobenzofuran-1,3-dione, 1,3-isobenzofurandione, 5-methyl, unii-2p2740fpck, phthalic anhydride, 4-methyl, dsstox_cid_7805, 5-methyl-1,3-dihydro-2-benzofuran-1,3-dione, dsstox_rid_78575, dsstox_gsid_27805, 4methylphthalic anhydride
PubChem CID 88069
IUPAC Name 5-methyl-2-benzofuran-1,3-dione
SMILES CC1=CC2=C(C=C1)C(=O)OC2=O

Specifications

CAS Min % 95.0
CAS Max % 100.0
Melting Point 88.0°C to 92.0°C
Infrared Spectrum Authentic
Assay Percent Range 95% min. (GC)
Packaging Glass bottle
Quantity 25 g
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in chloroform
Formula Weight 162.14
Percent Purity 96%
Physical Form Crystalline Powder or Flakes
Chemical Name or Material 4-Methylphthalic anhydride
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 243-073-6

RUO – Research Use Only