missing translation for 'onlineSavingsMsg'
Learn More

4-Methylphenylzinc iodide, 0.5M in THF, packaged under Argon in resealable ChemSeal™ bottles

Catalog No. aah58338ad
Change view
Click to view available options
Quantity:
50 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH58338AD 50 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH58338AD Supplier Thermo Scientific Chemicals Supplier No. H58338AD
Only null left

CAS: 300766-53-4 | C7H7IZn | 283.417 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 300766-53-4
Molecular Formula C7H7IZn
Molecular Weight (g/mol) 283.417
MDL Number MFCD01311454
InChI Key AHAJMZHMDYRVRB-UHFFFAOYSA-M
Synonym zinc 2+ ion 4-methylbenzen-1-ide iodide
PubChem CID 54719946
IUPAC Name iodozinc(1+);methylbenzene
SMILES CC1=CC=[C-]C=C1.[Zn+]I

Specifications

Density 0.997
Quantity 50 mL
UN Number UN3399
Beilstein 9694840
Sensitivity Air and light sensitive
Formula Weight 283.42
Chemical Name or Material 4-Methylphenylzinc iodide, Packaged under Argon in resealable ChemSeal™ bottles
Hazard Category H225-H302-H314-H335-H336-H351
Hazard Statement GHS H Statement
H225-H261-H314-H351-H335
Highly flammable liquid and vapor.
In contact with water releases flammable gas.
Causes severe skin burns and eye damage.
Suspected of causing cancer.
May cause respiratory irritation.
Precautionary Statement P201-P202-P210-P233-P235-P240-P241-P242-P243-P260-P264b-P270-P271-P281-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 4.3; Packing Group: II; Proper Shipping Name: ORGANOMETALLIC SUBSTANCE, LIQUID, WATER-REACTIVE, FLAMMABLE
TSCA No
Recommended Storage Keep cold; Store under Argon

RUO – Research Use Only