missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Methylphenylboronic Acid (contains varying amounts of Anhydride), TCI America™
$82.79 - $82.79
Chemical Identifiers
| CAS | 5720-05-8 |
|---|---|
| Molecular Formula | C7H9BO2 |
| Molecular Weight (g/mol) | 135.96 |
| MDL Number | MFCD00039138 |
| InChI Key | BIWQNIMLAISTBV-UHFFFAOYSA-N |
| PubChem CID | 79799 |
| IUPAC Name | (4-methylphenyl)boronic acid |
Chemical Identifiers
| 5720-05-8 | |
| 135.96 | |
| BIWQNIMLAISTBV-UHFFFAOYSA-N | |
| (4-methylphenyl)boronic acid |
| C7H9BO2 | |
| MFCD00039138 | |
| 79799 |
Specifications
| 5720-05-8 | |
| MFCD00039138 | |
| 4-tolylboronic acid, p-tolylboronic acid, 4-methylphenyl boronic acid, p-tolueneboronic acid, 4-methylbenzeneboronic acid, p-methylbenzeneboronic acid, boronic acid, 4-methylphenyl, boronic acid, p-tolyl, 4-methylphenylboric acid, tolyl boronic acid | |
| CC1=CC=C(C=C1)B(O)O | |
| 135.96 | |
| 135.96 | |
| 4-Methylphenylboronic Acid (contains varying amounts of Anhydride) |
| C7H9BO2 | |
| 5 g | |
| BIWQNIMLAISTBV-UHFFFAOYSA-N | |
| (4-methylphenyl)boronic acid | |
| 79799 | |
| Crystalline Powder |
Safety and Handling
RTECSNumber : XS7400000
TSCA : No