missing translation for 'onlineSavingsMsg'
Learn More

4-Methylphenylboronic Acid (contains varying amounts of Anhydride), TCI America™

Catalog No. M11265G
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M11265G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M11265G Supplier TCI America Supplier No. M11265G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 5720-05-8
Molecular Formula C7H9BO2
Molecular Weight (g/mol) 135.96
MDL Number MFCD00039138
InChI Key BIWQNIMLAISTBV-UHFFFAOYSA-N
PubChem CID 79799
IUPAC Name (4-methylphenyl)boronic acid

Specifications

Quantity 5 g
Synonym 4-tolylboronic acid, p-tolylboronic acid, 4-methylphenyl boronic acid, p-tolueneboronic acid, 4-methylbenzeneboronic acid, p-methylbenzeneboronic acid, boronic acid, 4-methylphenyl, boronic acid, p-tolyl, 4-methylphenylboric acid, tolyl boronic acid
SMILES CC1=CC=C(C=C1)B(O)O
Formula Weight 135.96
Physical Form Crystalline Powder
Chemical Name or Material 4-Methylphenylboronic Acid (contains varying amounts of Anhydride)
RTECSNumber XS7400000
TSCA No